APO455836684 Apollo Scientific Ltd

Methyl 4-fluoro-2-(methoxymethyl)-5-nitrobenzoate

  • MDL number MFCD22378816
  • Cas Number : 1243164-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-5-(methoxycarbonyl)-4-(methoxymethyl)nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50919

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50919
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart