APO455836685 Apollo Scientific Ltd

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid

  • MDL number MFCD00206674
  • Cas Number : 223499-20-5
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 212-215
Iupac Name 5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]-2-thiophenecarboxylic acid
Inchi Code 1S/C10H7F3N2O2S/c1-15-8(10(11,12)13)4-5(14-15)6-2-3-7(18-6)9(16)17/h2-4H,1H3,(H,16,17)
Inchi Key MGYVPTDPOYVOQK-UHFFFAOYSA-N

Description

Synonym 3-(5-Carboxythien-2-yl)-1-methyl-5-(trifluoromethyl)-1H-pyrazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 212-215
Product Link https://store.apolloscientific.co.uk/product/5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-ylthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-ylthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart