APO455836690 Apollo Scientific Ltd

4-Bromo-2,3-difluoroaniline

  • MDL number MFCD09030643
  • Cas Number : 112279-72-8
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-23-difluoroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-23-difluoroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart