APO455836694 Apollo Scientific Ltd

5-Bromo-3-fluoro-N1-methylbenzene-1,2-diamine

  • MDL number MFCD22415278
  • Cas Number : 1396503-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-fluoro-6-(methylamino)aniline, 5-Bromo-3-fluoro-N1-methylphenylene-1,2-diamine
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50931

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50931
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart