APO455836696 Apollo Scientific Ltd

2,3-Difluoro-4-iodobenzaldehyde

  • MDL number MFCD09835190
  • Cas Number : 885590-99-8
  • Molecular Weight : 268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-iodobenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart