APO455836699 Apollo Scientific Ltd

3-Hydroxy-5-(trifluoromethyl)benzaldehyde

  • MDL number MFCD16999720
  • Cas Number : 1243312-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Formyl-5-(trifluoromethyl)phenol, 3-Formyl-5-hydroxybenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51153

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51153
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart