APO455836707 Apollo Scientific Ltd

4-Bromo-2-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD18447612
  • Cas Number : 1394346-22-5
  • Molecular Weight : 268.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Borono-5-bromobenzotrifluoride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-trifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart