APO455836711 Apollo Scientific Ltd

5-Bromo-2-(trifluoromethoxy)benzeneboronic acid

  • MDL number MFCD11504857
  • Cas Number : 1072951-56-4
  • Molecular Weight : 284.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-trifluoromethoxybenzeneboronic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart