APO455836712 Apollo Scientific Ltd

Methyl 2-fluoro-3-nitrobenzoate

  • MDL number MFCD10566813
  • Cas Number : 946126-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-3-(methoxycarbonyl)nitrobenzene
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-2-fluoro-3-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-fluoro-3-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart