APO455836717 Apollo Scientific Ltd

5-(Pentafluorothio)-1,3-benzoxazole

  • MDL number MFCD22370119
  • Cas Number : 1379812-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1,3-Benzoxazol-5-yl)sulphur pentafluoride, 5-(Pentafluorosulphanyl)benzo[d]oxazole
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51185

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51185
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart