APO455836718 Apollo Scientific Ltd

5-(Pentafluorothio)-1H-benzimidazole

  • MDL number MFCD22370117
  • Cas Number : 663179-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1H-Benzimidazol-5-yl)sulphur pentafluoride, 5-(Pentafluorosulphanyl)-1H-benzimidazole
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51184

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=51184
Availability details for
Quantity
  • Availability
  • Add to Cart