APO455836722 Apollo Scientific Ltd

4-Hydroxy-2-(trifluoromethyl)pyridine

  • MDL number MFCD01571202
  • Cas Number : 170886-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Trifluoromethyl)pyridin-4-ol, 4-Hydroxy-alpha,alpha,alpha-trifluoro-2-picoline
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart