APO455836724 Apollo Scientific Ltd

4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine

  • MDL number MFCD11100737
  • Cas Number : 131747-61-0
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(Trifluoromethyl)pyridin-4-yl]methanol, 2-(Trifluoromethyl)isonicotinyl alcohol
Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxymethyl-2-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxymethyl-2-trifluoromethylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart