APO455836735 Apollo Scientific Ltd

(2-Fluorobenzyl)hydrazine

  • MDL number MFCD07785903
  • Cas Number : 51859-98-4
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Fluoro-2-(hydrazinomethyl)benzene
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluorobenzylhydrazine

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluorobenzylhydrazine
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart