APO455836742 Apollo Scientific Ltd

3-Nitro-4-(trifluoromethyl)benzyl bromide

  • MDL number MFCD16606359
  • Cas Number : 1227572-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-2-nitro-1-(trifluoromethyl)benzene, 4-(Bromomethyl)-2-nitrobenzotrifluoride
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=52994

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=52994
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart