APO455836745 Apollo Scientific Ltd

Methyl 2-fluoro-4-iodonicotinate

  • MDL number MFCD04972401
  • Cas Number : 884494-84-2
  • Molecular Weight : 281.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 2-fluoro-4-iodopyridine-3-carboxylate, 2-Fluoro-4-iodo-3-(methoxycarbonyl)pyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-fluoro-4-iodonicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-fluoro-4-iodonicotinate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart