APO455836749 Apollo Scientific Ltd

2-Bromo-6-nitro-4-[(trifluoromethyl)sulphonyl]phenol

  • MDL number MFCD24850030
  • Cas Number : 1440535-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-2-hydroxy-5-[(trifluoromethyl)sulphonyl]nitrobenzene, 3-Bromo-4-hydroxy-5-nitrophenyl trifluoromethyl sulphone
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53070

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53070
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart