APO455836752 Apollo Scientific Ltd

2,4-Difluoro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD24850039
  • Cas Number : 1440535-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Cyano-2,6-difluorobenzotrifluoride, alpha,alpha,alpha,2,4-Pentafluoro-m-tolunitrile
Purity 96
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53153

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53153
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart