APO455836761 Apollo Scientific Ltd

4-Fluoro-3-iodobenzyl bromide

  • MDL number MFCD08236849
  • Cas Number : 260050-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-1-fluoro-2-iodobenzene, alpha-Bromo-4-fluoro-3-iodotoluene
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=56894

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=56894
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart