APO455836767 Apollo Scientific Ltd

2,2,2-Trifluoro-2'-(trifluoromethyl)acetophenone

  • MDL number MFCD07380778
  • Cas Number : 42452-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2,2-Trifluoro-1-[2-(trifluoromethyl)phenyl]ethan-1-one, 2-(Trifluoroacetyl)benzotrifluoride
Purity 96
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=60327

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=60327
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart