APO455836769 Apollo Scientific Ltd

4'-Iodo-2'-(trifluoromethyl)acetanilide

  • MDL number MFCD00052845
  • Cas Number : 97760-98-0
  • Molecular Weight : 329.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-136
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-2-trifluoromethylacetanilide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodo-2-trifluoromethylacetanilide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart