APO455836772 Apollo Scientific Ltd

3-Chloro-2-iodo-2-(trifluoromethyl)-1,1,1-trifluoropropane

  • MDL number MFCD02093304
  • Cas Number : 240122-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Chloromethyl)-2-iodohexafluoropropane, 3-Chloro-2-(trifluoromethyl)-2-iodo-1,1,1-trifluoropropane
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 124-125
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2769

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2769
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart