APO455836773 Apollo Scientific Ltd

2-Bromo-4,5-difluorobenzyl alcohol

  • MDL number MFCD18909667
  • Cas Number : 476620-55-0
  • Molecular Weight : 223.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Synonym (2-Bromo-4,5-difluorophenyl)methanol
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-45-difluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-45-difluorobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart