APO455836774 Apollo Scientific Ltd

2-Bromo-4,5-difluorobenzyl bromide

  • MDL number MFCD18909668
  • Cas Number : 647862-95-1
  • Molecular Weight : 285.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
Iupac Name 1-bromo-2-(bromomethyl)-4,5-difluorobenzene
Inchi Code 1S/C7H4Br2F2/c8-3-4-1-6(10)7(11)2-5(4)9/h1-2H,3H2
Inchi Key GGZAHPYULZNIHT-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 1-Bromo-2-(bromomethyl)-4,5-difluorobenzene, alpha,2-Dibromo-4,5-difluorotoluene
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-45-difluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-45-difluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart