APO455836775 Apollo Scientific Ltd

4-(Trifluoroacetyl)piperidine hydrochloride

  • MDL number MFCD22041637
  • Cas Number : 1372103-92-8
  • Molecular Weight : 217.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Piperidin-4-yl)-2,2,2-trifluoroethan-1-one hydrochloride, 1-Oxo-1-(piperidin-4-yl)-2,2,2-trifluoroethane hydrochloride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-trifluoroacetylpiperidine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-trifluoroacetylpiperidine-hydrochloride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart