APO455836789 Apollo Scientific Ltd

Isopropyl 4,4,4-trifluoroacetoacetate

  • MDL number MFCD00040990
  • Cas Number : 175230-50-9
  • Molecular Weight : 198.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42/28mm
Product Link https://store.apolloscientific.co.uk/product/isopropyl-444-trifluoroacetoacetate

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/isopropyl-444-trifluoroacetoacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart