APO455836790 Apollo Scientific Ltd

3-Bromo-5-fluoronitrobenzene

  • MDL number MFCD09955454
  • Cas Number : 7087-65-2
  • Molecular Weight : 220

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 27-29
Synonym 1-Bromo-3-fluoro-5-nitrobenzene
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 65-70
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-fluoronitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-fluoronitrobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart