APO455836791 Apollo Scientific Ltd

3-Amino-6-bromo-2-fluorobenzotrifluoride

  • MDL number MFCD20483444
  • Cas Number : 159329-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-fluoro-3-(trifluoromethyl)aniline, 4-Bromo-alpha,alpha,alpha,2-tetrafluoro-m-toluidine
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53328

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53328
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart