APO455836796 Apollo Scientific Ltd

6-Bromo-2-hydroxy-3-nitrobenzotrifluoride

  • MDL number MFCD25541885
  • Cas Number : 1445995-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-6-nitro-2-(trifluoromethyl)phenol, 3-Bromo-6-nitro-alpha,alpha,alpha-trifluoro-o-cresol
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54163

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54163
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart