APO455836799 Apollo Scientific Ltd

5-Bromo-2-methoxy-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD25541888
  • Cas Number : 1445995-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-(trifluoromethyl)-o-anisonitrile, 5-Bromo-3-cyano-2-methoxybenzotrifluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54683

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54683
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart