APO455836802 Apollo Scientific Ltd

2-Hydroxy-3-(trifluoromethyl)benzoic acid

  • MDL number MFCD06203614
  • Cas Number : 251300-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-163
Synonym 3-(Trifluoromethyl)salicylic acid, 3-Carboxy-2-hydroxybenzotrifluoride
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54689

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54689
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart