APO455836804 Apollo Scientific Ltd

3-(Pentafluorothio)benzyl alcohol

  • MDL number MFCD06203062
  • Cas Number : 773872-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Hydroxymethyl)sulphur pentafluoride, 3-(Pentafluorosulphanyl)benzyl alcohol
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54693

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54693
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart