APO455836807 Apollo Scientific Ltd

4-Fluoro-2-nitrothiophenol

  • MDL number MFCD22481385
  • Cas Number : 654063-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoro-2-nitrobenzenethiol, 4-Fluoro-2-nitrophenyl mercaptan
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54696

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54696
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart