APO455836809 Apollo Scientific Ltd

5-Iodo-2-methoxy-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD25541890
  • Cas Number : 1445995-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Iodo-3-(trifluoromethyl)-o-anisonitrile, 3-Cyano-5-iodo-2-methoxybenzotrifluoride
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=54851

Safety Information

signalword Danger
pcodes P312;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=54851
pictograms GHS06;GHS07
hstatements H302+H332;H311;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart