APO455836811 Apollo Scientific Ltd

Methyl 2-bromo-4,5-difluorobenzoate

  • MDL number MFCD08458104
  • Cas Number : 878207-28-4
  • Molecular Weight : 251.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 28-29
Iupac Name methyl 2-bromo-4,5-difluorobenzoate
Inchi Code 1S/C8H5BrF2O2/c1-13-8(12)4-2-6(10)7(11)3-5(4)9/h2-3H,1H3
Inchi Key ZCGAVEFBZYWYJW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 28-29
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-45-difluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-45-difluorobenzoate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart