APO455836814 Apollo Scientific Ltd

Methyl 4-hydroxy-2,3,5-trifluorobenzoate

  • MDL number MFCD14698055
  • Cas Number : 1214387-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-131
Synonym 4-(Methoxycarbonyl)-2,3,6-trifluorophenol
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63384

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=63384
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart