APO455836815 Apollo Scientific Ltd

4-Hydroxy-2,3,5-trifluorobenzoic acid

  • MDL number MFCD08458460
  • Cas Number : 156839-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-161
Synonym 4-Carboxy-2,3,6-trifluorophenol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63385

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63385
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart