APO455836823 Apollo Scientific Ltd

3-Bromo-5-hydroxybenzotrifluoride

  • MDL number MFCD11041083
  • Cas Number : 1025718-84-6
  • Molecular Weight : 241.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-5-(trifluoromethyl)phenol, 5-Bromo-alpha,alpha,alpha-trifluoro-m-cresol
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-5-hydroxybenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-5-hydroxybenzotrifluoride
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart