APO455836826 Apollo Scientific Ltd

6-(Trifluoromethyl)isatoic anhydride

  • MDL number MFCD24850066
  • Cas Number : 387-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239
Synonym 5-(Trifluoromethyl)-2H-3,1-benzoxazine-2,4(1H)-dione, 5-(Trifluoromethyl)-1H-benzo[d][1,3]oxazine-2,4-dione
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53315

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53315
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart