APO455836827 Apollo Scientific Ltd

4,5-Difluoroisatoic anhydride

  • MDL number MFCD08457975
  • Cas Number : 97927-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223-226
Synonym 6,7-Difluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 6,7-Difluoro-1H-benzo[d][1,3]oxazine-2,4-dione
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53319

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53319
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart