APO455836829 Apollo Scientific Ltd

2-Chloro-6-fluorophenyl trifluoromethanesulphonate

  • MDL number MFCD25541813
  • Cas Number : 1443684-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-fluorophenyl triflate
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55132

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55132
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart