APO455836832 Apollo Scientific Ltd

3-Bromobenzal fluoride

  • MDL number MFCD07368984
  • Cas Number : 29848-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-(difluoromethyl)benzene, 3-Bromo-alpha,alpha-difluorotoluene
Purity 97
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48023

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48023
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart