APO455836842 Apollo Scientific Ltd

4-Nitro-3-(trifluoromethyl)phenyl trifluoromethanesulphonate

  • MDL number MFCD25541875
  • Cas Number : 1446016-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Nitro-3-(trifluoromethyl)phenyl triflate, 2-Nitro-5-{[(trifluoromethyl)sulphonyl]oxy}benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55196

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55196
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart