APO455836843 Apollo Scientific Ltd

2-Nitro-4-(trifluoromethoxy)phenyl trifluoromethanesulphonate

  • MDL number MFCD25541876
  • Cas Number : 955395-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Nitro-4-(trifluoromethoxy)phenyl triflate, 5-(Trifluoromethoxy)-2-{[(trifluoromethyl)sulphonyl]oxy}nitrobenzene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55197

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55197
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart