APO455836846 Apollo Scientific Ltd

3'-Chloro-5'-(trifluoromethoxy)acetophenone

  • MDL number MFCD06660315
  • Cas Number : 886503-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[3-Chloro-5-(trifluoromethoxy)phenyl]ethan-1-one
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=49476

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=49476
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart