APO455836847 Apollo Scientific Ltd

2-Bromo-4-fluoro-5-nitrophenol

  • MDL number MFCD08460104
  • Cas Number : 84478-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Synonym 4-Bromo-2-fluoro-5-hydroxynitrobenzene
Purity 99
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=43193

Safety Information

signalword Warning
pcodes P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=43193
pictograms GHS07
hstatements H302;H312;H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart