APO455836849 Apollo Scientific Ltd

2,3-Difluoro-4-iodobenzoic acid

  • MDL number MFCD07368123
  • Cas Number : 501433-05-2
  • Molecular Weight : 284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-197
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-iodobenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart