APO455836850 Apollo Scientific Ltd

3-Bromo-6-chloro-2-fluorobenzyl bromide

  • MDL number MFCD27931488
  • Cas Number : 886615-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-(bromomethyl)-4-chloro-2-fluorobenzene, 6-Chloro-alpha,3-dibromo-2-fluorotoluene
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=63432

Safety Information

signalword Danger
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=63432
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart