APO455836851 Apollo Scientific Ltd

3-Bromo-6-chloro-2-fluorobenzyl alcohol

  • MDL number MFCD18447606
  • Cas Number : 886615-31-2
  • Molecular Weight : 239.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Bromo-6-chloro-2-fluorophenyl)methanol
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-6-chloro-2-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-6-chloro-2-fluorobenzyl-alcohol
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart