APO455836853 Apollo Scientific Ltd

Methyl 2-(bromomethyl)-4-fluorobenzoate

  • MDL number MFCD11505951
  • Cas Number : 157652-28-3
  • Molecular Weight : 247.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methoxycarbonyl)-5-fluorobenzyl bromide, Methyl alpha-bromo-4-fluoro-o-toluate
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromomethyl-4-fluorobenzoate

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromomethyl-4-fluorobenzoate
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart